The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024778.5 | NP_079054.3 | 2 - 3 | 1102 | 113 |
XM_017029845.1 | XP_016885334.1 | 2 - 3 | 1102 | 113 | |
GenBank mRNA | AK026265.1 | - | 2 - 3 | 354 | 113 |
AK290198.1 | - | 2 - 3 | 1098 | 113 | |
BC103491.1 | - | 2 - 3 | 966 | 113 |