The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031855.2 | NP_001027026.1 | 5 - 6 | 1588 | 62 |
NM_001289109.1 | NP_001276038.1 | 6 - 7 | 1374 | 62 | |
NM_024778.5 | NP_079054.3 | 4 - 5 | 1465 | 62 | |
NR_110311.1 | 5 - 6 | 1588 | 62 | ||
XM_005262476.2 | XP_005262533.1 | 6 - 7 | 1744 | 62 | |
XM_011531398.2 | XP_011529700.1 | 6 - 7 | 1744 | 62 | |
XM_017029845.1 | XP_016885334.1 | 4 - 5 | 1465 | 62 | |
GenBank mRNA | AK026265.1 | - | 4 - 5 | 717 | 62 |
AK091777.1 | - | 5 - 6 | 1584 | 62 | |
AK097641.1 | - | 6 - 7 | 1374 | 62 | |
AK290198.1 | - | 4 - 5 | 1461 | 62 | |
BC099847.1 | - | 5 - 6 | 1452 | 62 | |
BC100671.1 | - | 5 - 6 | 1452 | 62 | |
BC103491.1 | - | 4 - 5 | 1329 | 62 |