The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042467.2 | NP_001035932.1 | 14 - 15 | 1937 | 94 |
NM_001281473.1 | NP_001268402.1 | 12 - 13 | 1661 | 94 | |
NM_001281474.1 | NP_001268403.1 | 13 - 14 | 1742 | 94 | |
NM_024101.6 | NP_077006.1 | 15 - 16 | 2021 | 94 | |
NR_104019.1 | 16 - 17 | 2210 | 94 | ||
XM_006712737.1 | XP_006712800.1 | 14 - 15 | 1869 | 94 | |
XM_006712739.1 | XP_006712802.1 | 14 - 15 | 1833 | 94 | |
XM_006712740.1 | XP_006712803.1 | 13 - 14 | 1785 | 94 | |
XM_011511812.1 | XP_011510114.1 | 12 - 13 | 3681 | 94 | |
GenBank mRNA | AK022207.1 | - | 14 - 15 | 1905 | 94 |
AK056867.1 | - | 11 - 12 | 1915 | 94 | |
AK094168.1 | - | 13 - 14 | 1742 | 94 | |
AK225381.1 | - | 12 - 13 | 1606 | 94 | |
AK292929.1 | - | 14 - 15 | 1905 | 94 | |
AK296745.1 | - | 16 - 17 | 2179 | 94 | |
AY358857.1 | - | 10 - 11 | 1759 | 94 | |
BC001653.2 | - | 15 - 16 | 1953 | 94 | |
BC051269.1 | - | 14 - 15 | 1879 | 94 |