The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105214.2 | NP_001098684.1 | 5 - 6 | 736 | 93 |
NM_001261832.1 | NP_001248761.1 | 5 - 6 | 736 | 93 | |
NM_001282272.1 | NP_001269201.1 | 4 - 5 | 489 | 93 | |
NM_004674.4 | NP_004665.2 | 5 - 6 | 635 | 93 | |
XM_005273682.1 | XP_005273739.1 | 5 - 6 | 1163 | 93 | |
XM_005273683.1 | XP_005273740.1 | 5 - 6 | 736 | 93 | |
XM_006716412.1 | XP_006716475.1 | 5 - 6 | 1163 | 93 | |
XM_006716413.2 | XP_006716476.1 | 5 - 6 | 601 | 93 | |
GenBank mRNA | AB020982.1 | - | 5 - 6 | 588 | 93 |
AF056717.1 | - | 5 - 6 | 590 | 93 | |
AF056718.1 | - | 5 - 6 | 581 | 93 | |
AK291750.1 | - | 5 - 6 | 599 | 93 | |
AK291938.1 | - | 5 - 6 | 607 | 93 | |
AK298483.1 | - | 4 - 5 | 432 | 93 | |
AK311336.1 | - | 5 - 6 | 513 | 93 | |
BC015936.2 | - | 5 - 6 | 603 | 93 |