The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105214.2 | NP_001098684.1 | 9 - 10 | 1109 | 58 |
NM_001261832.1 | NP_001248761.1 | 9 - 10 | 1109 | 58 | |
NM_001282272.1 | NP_001269201.1 | 8 - 9 | 862 | 58 | |
NM_004674.4 | NP_004665.2 | 9 - 10 | 1008 | 58 | |
XM_005273682.1 | XP_005273739.1 | 10 - 11 | 1554 | 58 | |
XM_005273683.1 | XP_005273740.1 | 10 - 11 | 1127 | 58 | |
XM_006716412.1 | XP_006716475.1 | 10 - 11 | 1554 | 58 | |
XM_006716413.2 | XP_006716476.1 | 9 - 10 | 974 | 58 | |
GenBank mRNA | AB020982.1 | - | 9 - 10 | 961 | 58 |
AF056717.1 | - | 9 - 10 | 963 | 58 | |
AF056718.1 | - | 9 - 10 | 954 | 58 | |
AK291750.1 | - | 9 - 10 | 972 | 58 | |
AK291938.1 | - | 9 - 10 | 980 | 58 | |
AK298483.1 | - | 8 - 9 | 805 | 58 | |
AK311336.1 | - | 9 - 10 | 886 | 58 | |
BC015936.2 | - | 9 - 10 | 976 | 58 |