Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NARS
Entrez Gene ID
4677
Gene Name
asparaginyl-tRNA synthetase
Gene Aliases
ASNRS, NARS1
Chromosome Location
Chr.18: 57600656 - 57621945 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004539.3 NP_004530.1 13 - 14 1742 75
XM_005266700.1 XP_005266757.1 13 - 14 1520 75
GenBank mRNA AJ000334.1 - 13 - 14 1587 75
AK129897.1 - 13 - 14 1544 75
AK222835.1 - 13 - 14 1589 75
AK297777.1 - 11 - 12 1173 75
BC001687.1 - 13 - 14 1591 75
D84273.1 - 13 - 14 1546 75
U79254.1 - 1 - 2 35 75

View Full Product Details