The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005030.5 | NP_005021.2 | 3 - 4 | 883 | 85 |
GenBank mRNA | AB209179.1 | - | 2 - 3 | 2742 | 85 |
AK303263.1 | - | 4 - 5 | 499 | 85 | |
AK308276.1 | - | 4 - 5 | 740 | 85 | |
AK313227.1 | - | 3 - 4 | 807 | 85 | |
BC002369.2 | - | 3 - 4 | 775 | 85 | |
BC003002.1 | - | 3 - 4 | 753 | 85 | |
BC014846.1 | - | 3 - 4 | 728 | 85 | |
L19559.1 | - | 3 - 4 | 785 | 85 | |
U01038.1 | - | 3 - 4 | 785 | 85 | |
X73458.1 | - | 3 - 4 | 799 | 85 | |
X75932.1 | - | 3 - 4 | 785 | 85 |