The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002755.3 | NP_002746.1 | 3 - 4 | 910 | 70 |
XM_011521783.2 | XP_011520085.1 | 3 - 4 | 567 | 70 | |
GenBank mRNA | AK291500.1 | - | 3 - 4 | 871 | 70 |
AK294322.1 | - | 3 - 4 | 421 | 70 | |
AK312356.1 | - | 3 - 4 | 817 | 70 | |
BC137459.1 | - | 3 - 4 | 498 | 70 | |
BC139729.1 | - | 3 - 4 | 494 | 70 | |
L05624.1 | - | 3 - 4 | 468 | 70 | |
L11284.1 | - | 3 - 4 | 507 | 70 |