The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002755.3 | NP_002746.1 | 5 - 6 | 1040 | 73 |
XM_011521783.2 | XP_011520085.1 | 5 - 6 | 697 | 73 | |
XM_017022411.1 | XP_016877900.1 | 4 - 5 | 913 | 73 | |
XM_017022412.1 | XP_016877901.1 | 4 - 5 | 499 | 73 | |
GenBank mRNA | AK291500.1 | - | 5 - 6 | 1001 | 73 |
AK294322.1 | - | 5 - 6 | 551 | 73 | |
AK312356.1 | - | 5 - 6 | 947 | 73 | |
BC137459.1 | - | 5 - 6 | 628 | 73 | |
BC139729.1 | - | 5 - 6 | 624 | 73 | |
L05624.1 | - | 5 - 6 | 598 | 73 | |
L11284.1 | - | 5 - 6 | 637 | 73 |