The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308171.1 | NP_001295100.1 | 10 - 11 | 1183 | 110 |
NM_005925.2 | NP_005916.2 | 10 - 11 | 1183 | 110 | |
XM_011526013.2 | XP_011524315.1 | 9 - 10 | 949 | 110 | |
XM_011526014.2 | XP_011524316.1 | 8 - 9 | 796 | 110 | |
GenBank mRNA | AY695931.1 | - | 10 - 11 | 1194 | 110 |
BC136559.1 | - | 10 - 11 | 1161 | 110 | |
BC144244.1 | - | 10 - 11 | 1161 | 110 | |
X81333.1 | - | 10 - 11 | 1182 | 110 |