Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALG6
Entrez Gene ID
29929
Gene Name
ALG6, alpha-1,3-glucosyltransferase
Gene Aliases
CDG1C
Chromosome Location
Chr.1: 63367590 - 63438562 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013339.3 NP_037471.2 13 - 14 1433 124
GenBank mRNA AF102851.1 - 13 - 14 1366 124
AK022700.1 - 13 - 14 1371 124
BC001253.1 - 13 - 14 1365 124
CB215801.1 - 5 - 6 359 124

View Full Product Details