Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADO
Entrez Gene ID
84890
Gene Name
2-aminoethanethiol dioxygenase
Gene Aliases
C10orf22
Chromosome Location
Chr.10: 62804756 - 62808479 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032804.5 NP_116193.2 1 - 1 257 157
GenBank mRNA AK127694.1 - 1 - 1 259 157
BC018660.1 - 1 - 1 161 157
BC028589.2 - 1 - 1 127 157

View Full Product Details