The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304964.1 | NP_001291893.1 | 1 - 2 | 257 | 86 |
NM_152779.3 | NP_689992.1 | 1 - 2 | 257 | 86 | |
XM_011538134.2 | XP_011536436.2 | 1 - 2 | 3052 | 86 | |
GenBank mRNA | AY358731.1 | - | 1 - 2 | 257 | 86 |
BC014603.1 | - | 1 - 2 | 218 | 86 |