Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGMS1
Entrez Gene ID
259230
Gene Name
sphingomyelin synthase 1
Gene Aliases
MOB, MOB1, SMS1, TMEM23, hmob33
Chromosome Location
Chr.10: 50305585 - 50623977 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_147156.3 NP_671512.1 5 - 6 642 97
XM_011539582.2 XP_011537884.1 2 - 3 246 97
XM_011539584.2 XP_011537886.1 2 - 3 246 97
GenBank mRNA AK026683.1 - 1 - 2 105 97
AK311737.1 - 5 - 6 658 97
AY280959.1 - 5 - 6 608 97
AY332650.1 - 5 - 6 517 97
AY364088.1 - 5 - 6 517 97
KP710931.1 - 4 - 5 331 97
KP710935.1 - - 74 97
KR020014.1 - 3 - 4 322 97
KR020022.1 - 5 - 6 558 97

View Full Product Details