Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGMS1
Entrez Gene ID
259230
Gene Name
sphingomyelin synthase 1
Gene Aliases
MOB, MOB1, SMS1, TMEM23, hmob33
Chromosome Location
Chr.10: 50305585 - 50623977 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_147156.3 NP_671512.1 7 - 8 1575 125
XM_005269675.1 XP_005269732.1 2 - 3 1267 125
XM_011539582.2 XP_011537884.1 4 - 5 1179 125
XM_011539583.2 XP_011537885.1 2 - 3 1056 125
GenBank mRNA AB154421.1 - 3 - 4 958 125
AK026683.1 - 3 - 4 1038 125
AY280959.1 - 7 - 8 1541 125
AY312431.1 - 1 - 2 767 125
AY332650.1 - 7 - 8 1450 125
BC042899.1 - 1 - 2 836 125
FN908146.1 - 2 - 3 969 125

View Full Product Details