The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166139.2 | NP_001159611.1 | 4 - 5 | 560 | 153 |
NM_153686.8 | NP_710153.2 | 4 - 5 | 560 | 153 | |
NR_136669.1 | 3 - 4 | 480 | 153 | ||
XM_011513821.2 | XP_011512123.1 | 4 - 5 | 438 | 153 | |
XM_011513822.2 | XP_011512124.1 | 4 - 5 | 438 | 153 | |
XM_011513823.2 | XP_011512125.1 | 3 - 4 | 284 | 153 | |
XM_011513825.2 | XP_011512127.1 | 4 - 5 | 438 | 153 | |
XM_011513826.2 | XP_011512128.1 | 4 - 5 | 438 | 153 | |
XM_017007962.1 | XP_016863451.1 | 3 - 4 | 362 | 153 | |
XM_017007963.1 | XP_016863452.1 | 3 - 4 | 665 | 153 | |
GenBank mRNA | AK096181.1 | - | 5 - 6 | 421 | 153 |
AK299939.1 | - | 3 - 4 | 362 | 153 | |
BC037322.1 | - | 4 - 5 | 443 | 153 | |
BU902997.1 | - | 5 - 6 | 569 | 153 | |
BX354708.2 | - | 3 - 4 | 153 |