The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256875.1 | NP_001243804.1 | 5 - 6 | 716 | 145 |
NM_018101.3 | NP_060571.1 | 6 - 7 | 619 | 145 | |
GenBank mRNA | AK001330.1 | - | 6 - 7 | 539 | 145 |
AK022104.1 | - | 5 - 6 | 621 | 145 | |
AK022606.1 | - | 5 - 6 | 525 | 145 | |
AK222549.1 | - | 6 - 7 | 539 | 145 | |
AK222568.1 | - | 6 - 7 | 493 | 145 | |
AK291917.1 | - | 5 - 6 | 571 | 145 | |
AK302116.1 | - | 4 - 5 | 383 | 145 | |
AY508815.2 | - | 6 - 7 | 539 | 145 | |
BC000703.2 | - | 5 - 6 | 612 | 145 | |
BC001651.2 | - | 5 - 6 | 630 | 145 | |
BC008079.2 | - | 6 - 7 | 472 | 145 | |
BC016944.1 | - | 6 - 7 | 492 | 145 |