The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003325.3 | NP_003316.3 | 2 - 3 | 319 | 64 |
GenBank mRNA | AK289809.1 | - | 2 - 3 | 99 | 64 |
AK299950.1 | - | 3 - 4 | 258 | 64 | |
AK302835.1 | - | 3 - 4 | 363 | 64 | |
AK308642.1 | - | 3 - 4 | 258 | 64 | |
BC032721.1 | - | 2 - 3 | 357 | 64 | |
BC039835.1 | - | 2 - 3 | 314 | 64 | |
CR456503.1 | - | 2 - 3 | 105 | 64 | |
X75296.1 | - | 2 - 3 | 326 | 64 | |
X77633.1 | - | 1 - 2 | 75 | 64 | |
X81844.1 | - | 2 - 3 | 160 | 64 | |
X89887.1 | - | 2 - 3 | 319 | 64 |