The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003105.5 | NP_003096.1 | 24 - 25 | 3590 | 80 |
XM_011542963.2 | XP_011541265.1 | 23 - 24 | 3351 | 80 | |
XM_011542965.2 | XP_011541267.1 | 18 - 19 | 2499 | 80 | |
XM_011542967.2 | XP_011541269.1 | 4 - 5 | 531 | 80 | |
XM_017018169.1 | XP_016873658.1 | 24 - 25 | 3297 | 80 | |
XM_017018170.1 | XP_016873659.1 | 22 - 23 | 2973 | 80 | |
XM_017018171.1 | XP_016873660.1 | 24 - 25 | 3465 | 80 | |
XM_017018172.1 | XP_016873661.1 | 10 - 11 | 1462 | 80 | |
GenBank mRNA | AK295878.1 | - | 4 - 5 | 531 | 80 |
BC137171.1 | - | 24 - 25 | 3513 | 80 | |
Y08110.1 | - | 24 - 25 | 3541 | 80 |