Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORL1
Entrez Gene ID
6653
Gene Name
sortilin related receptor 1
Gene Aliases
C11orf32, LR11, LRP9, SORLA, SorLA-1, gp250
Chromosome Location
Chr.11: 121452203 - 121633762 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003105.5 NP_003096.1 2 - 3 531 118
XM_011542963.2 XP_011541265.1 2 - 3 406 118
XM_017018169.1 XP_016873658.1 2 - 3 238 118
XM_017018171.1 XP_016873660.1 2 - 3 406 118
GenBank mRNA AK309523.1 - 2 - 3 354 118
BC137171.1 - 2 - 3 454 118
BX365721.2 - 2 - 3 531 118
DN831521.1 - 1 - 2 93 118
Y08110.1 - 2 - 3 482 118

View Full Product Details