The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003105.5 | NP_003096.1 | 42 - 43 | 5854 | 101 |
XM_011542963.2 | XP_011541265.1 | 41 - 42 | 5615 | 101 | |
XM_011542965.2 | XP_011541267.1 | 36 - 37 | 4763 | 101 | |
XM_011542967.2 | XP_011541269.1 | 22 - 23 | 2795 | 101 | |
XM_017018169.1 | XP_016873658.1 | 42 - 43 | 5561 | 101 | |
XM_017018170.1 | XP_016873659.1 | 40 - 41 | 5237 | 101 | |
XM_017018172.1 | XP_016873661.1 | 28 - 29 | 3726 | 101 | |
GenBank mRNA | AK294404.1 | - | 13 - 14 | 1992 | 101 |
AK295090.1 | - | 19 - 20 | 2644 | 101 | |
AK295878.1 | - | 22 - 23 | 2795 | 101 | |
AK302589.1 | - | 19 - 20 | 2466 | 101 | |
AK316340.1 | - | 13 - 14 | 1574 | 101 | |
BC137171.1 | - | 42 - 43 | 5777 | 101 | |
Y08110.1 | - | 42 - 43 | 5805 | 101 |