The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003105.5 | NP_003096.1 | 7 - 8 | 1173 | 86 |
XM_011542963.2 | XP_011541265.1 | 7 - 8 | 1048 | 86 | |
XM_017018169.1 | XP_016873658.1 | 7 - 8 | 880 | 86 | |
XM_017018170.1 | XP_016873659.1 | 5 - 6 | 556 | 86 | |
XM_017018171.1 | XP_016873660.1 | 7 - 8 | 1048 | 86 | |
GenBank mRNA | AK309523.1 | - | 7 - 8 | 996 | 86 |
BC137171.1 | - | 7 - 8 | 1096 | 86 | |
Y08110.1 | - | 7 - 8 | 1124 | 86 |