Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NTRK3
Entrez Gene ID
4916
Gene Name
neurotrophic receptor tyrosine kinase 3
Gene Aliases
GP145-TrkC, TRKC, gp145(trkC)
Chromosome Location
Chr.15: 87859749 - 88256796 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007156.2 NP_001007157.1 8 - 9 1068 60
NM_001012338.2 NP_001012338.1 8 - 9 1068 60
NM_001243101.1 NP_001230030.1 8 - 9 1068 60
NM_001320134.1 NP_001307063.1 6 - 7 778 60
NM_001320135.1 NP_001307064.1 6 - 7 669 60
NM_002530.3 NP_002521.2 8 - 9 1068 60
XM_006720543.3 XP_006720606.1 6 - 7 1377 60
XM_006720544.3 XP_006720607.1 6 - 7 1377 60
XM_006720545.3 XP_006720608.1 6 - 7 1377 60
XM_006720548.3 XP_006720611.1 6 - 7 1377 60
XM_006720549.3 XP_006720612.1 6 - 7 1377 60
XM_006720550.3 XP_006720613.1 6 - 7 1377 60
XM_011521637.2 XP_011519939.1 6 - 7 1377 60
XM_011521638.2 XP_011519940.1 6 - 7 1377 60
XM_017022240.1 XP_016877729.1 7 - 8 1248 60
XM_017022241.1 XP_016877730.1 6 - 7 733 60
XM_017022242.1 XP_016877731.1 6 - 7 1377 60
XM_017022243.1 XP_016877732.1 6 - 7 729 60
XM_017022244.1 XP_016877733.1 6 - 7 631 60
XM_017022245.1 XP_016877734.1 7 - 8 686 60
XM_017022246.1 XP_016877735.1 6 - 7 787 60
XM_017022247.1 XP_016877736.1 6 - 7 1377 60
XM_017022248.1 XP_016877737.1 6 - 7 1377 60
XM_017022249.1 XP_016877738.1 6 - 7 1377 60
XM_017022250.1 XP_016877739.1 6 - 7 1377 60
XM_017022253.1 XP_016877742.1 6 - 7 1377 60
XM_017022254.1 XP_016877743.1 6 - 7 1377 60
GenBank mRNA AK297160.1 - 6 - 7 786 60
AK302504.1 - 6 - 7 628 60
AK309695.1 - 6 - 7 788 60
AK313646.1 - 6 - 7 787 60
BC013693.1 - 6 - 7 767 60
BC128249.1 - 2 - 3 224 60
BT007291.1 - 6 - 7 762 60
KJ534904.1 - 6 - 7 762 60
KJ534905.1 - 6 - 7 762 60
KJ534906.1 - 6 - 7 762 60
KJ535052.1 - 6 - 7 762 60
S76475.1 - 6 - 7 777 60
S76476.1 - 6 - 7 777 60
U05012.1 - 7 - 8 917 60

View Full Product Details