Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NTRK3
- Entrez Gene ID
-
4916
- Gene Name
- neurotrophic receptor tyrosine kinase 3
- Gene Aliases
-
GP145-TrkC, TRKC, gp145(trkC)
- Chromosome Location
-
Chr.15: 87859749 - 88256796 on Build GRCh38
View Full Product Details