The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001310319.1 | NP_001297248.1 | 6 - 7 | 1042 | 86 |
NM_001310320.1 | NP_001297249.1 | 8 - 9 | 1227 | 86 | |
NM_001310321.1 | NP_001297250.1 | 7 - 8 | 1286 | 86 | |
NM_005928.3 | NP_005919.2 | 7 - 8 | 1174 | 86 | |
GenBank mRNA | AK095908.1 | - | 8 - 9 | 1137 | 86 |
AK222735.1 | - | 7 - 8 | 1084 | 86 | |
AK223182.1 | - | 7 - 8 | 1084 | 86 | |
AK304627.1 | - | 6 - 7 | 952 | 86 | |
AK312640.1 | - | 7 - 8 | 1084 | 86 | |
AY141974.1 | - | 7 - 8 | 1093 | 86 | |
S56151.1 | - | 4 - 5 | 517 | 86 | |
U58516.1 | - | 7 - 8 | 1084 | 86 |