The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032726.2 | NP_001027898.2 | 5 - 6 | 135 | |
XM_011530948.2 | XP_011529250.1 | 5 - 6 | 135 | ||
XM_017029488.1 | XP_016884977.1 | 5 - 6 | 135 | ||
GenBank mRNA | AI126463.1 | - | 3 - 4 | 571 | 135 |
BC117239.1 | - | 5 - 6 | 135 |