The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032726.2 | NP_001027898.2 | 7 - 8 | 91 | |
XM_017029488.1 | XP_016884977.1 | 7 - 8 | 91 | ||
GenBank mRNA | AI126463.1 | - | 1 - 2 | 427 | 91 |
BC117239.1 | - | 7 - 8 | 91 |