Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CCNYL2
- Entrez Gene ID
-
414194
- Gene Name
- cyclin Y-like 2 (pseudogene)
- Gene Aliases
-
C10orf21, CCNYL2P, bA178A10.2
- Chromosome Location
-
Chr.10: 42408174 - 42472240 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_103829.1 |
|
7 - 8 |
1125 |
140 |
View Full Product Details