Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCM3AP-AS1
Entrez Gene ID
114044
Gene Name
MCM3AP antisense RNA 1
Gene Aliases
C21orf85, MCM3AP-AS, MCM3APAS, MCM3APASB, NCRNA00031
Chromosome Location
Chr.21: 46229231 - 46251701 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_002776.4 2 - 3 215 134
NR_110565.1 3 - 4 416 134

View Full Product Details