Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JUP
Entrez Gene ID
3728
Gene Name
junction plakoglobin
Gene Aliases
ARVD12, CTNNG, DP3, DPIII, PDGB, PKGB
Chromosome Location
Chr.17: 41754607 - 41786853 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002230.2 NP_002221.1 6 - 7 1190 77
NM_021991.2 NP_068831.1 6 - 7 1190 77
XM_006721873.2 XP_006721936.1 7 - 8 1440 77
XM_006721874.2 XP_006721937.1 6 - 7 1353 77
XM_006721875.1 XP_006721938.1 6 - 7 1272 77
XM_006721878.1 XP_006721941.1 6 - 7 1253 77
XM_011524753.2 XP_011523055.1 7 - 8 1476 77
XM_011524755.1 XP_011523057.1 6 - 7 1185 77
XM_011524756.1 XP_011523058.1 6 - 7 1083 77
XM_011524757.2 XP_011523059.1 6 - 7 1350 77
XM_011524758.1 XP_011523060.1 6 - 7 1079 77
XM_017024588.1 XP_016880077.1 6 - 7 1172 77
XM_017024589.1 XP_016880078.1 6 - 7 1173 77
XM_017024590.1 XP_016880079.1 6 - 7 1079 77
GenBank mRNA AK226051.1 - 6 - 7 1167 77
AY243535.1 - 5 - 6 1063 77
BC000441.2 - 6 - 7 1176 77
BC011865.2 - 6 - 7 1334 77
KU178155.1 - 5 - 6 1057 77
M23410.1 - 6 - 7 1173 77
Z68228.1 - 6 - 7 1176 77

View Full Product Details