Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JUP
Entrez Gene ID
3728
Gene Name
junction plakoglobin
Gene Aliases
ARVD12, CTNNG, DP3, DPIII, PDGB, PKGB
Chromosome Location
Chr.17: 41754607 - 41786853 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002230.2 NP_002221.1 9 - 10 1786 62
NM_021991.2 NP_068831.1 9 - 10 1786 62
XM_006721873.2 XP_006721936.1 10 - 11 2036 62
XM_006721874.2 XP_006721937.1 9 - 10 1949 62
XM_006721875.1 XP_006721938.1 9 - 10 1868 62
XM_006721878.1 XP_006721941.1 9 - 10 1849 62
XM_011524753.2 XP_011523055.1 10 - 11 2072 62
XM_011524755.1 XP_011523057.1 9 - 10 1781 62
XM_011524756.1 XP_011523058.1 9 - 10 1679 62
XM_011524757.2 XP_011523059.1 9 - 10 1946 62
XM_011524758.1 XP_011523060.1 9 - 10 1675 62
XM_017024588.1 XP_016880077.1 9 - 10 1768 62
XM_017024589.1 XP_016880078.1 9 - 10 1769 62
XM_017024590.1 XP_016880079.1 9 - 10 1675 62
GenBank mRNA AK226051.1 - 9 - 10 1763 62
AY243535.1 - 8 - 9 1659 62
BC000441.2 - 9 - 10 1772 62
BC011865.2 - 9 - 10 1930 62
KU178155.1 - 8 - 9 1653 62
KU178156.1 - 3 - 4 420 62
M23410.1 - 9 - 10 1769 62
Z68228.1 - 9 - 10 1772 62

View Full Product Details