The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267059.1 | NP_001253988.1 | 7 - 8 | 817 | 72 |
NM_001267060.1 | NP_001253989.1 | 6 - 7 | 778 | 72 | |
NM_001267061.1 | NP_001253990.1 | 7 - 8 | 688 | 72 | |
NM_014748.3 | NP_055563.1 | 7 - 8 | 853 | 72 | |
NR_049782.1 | 7 - 8 | 978 | 72 | ||
NR_049783.1 | 8 - 9 | 951 | 72 | ||
NR_049784.1 | 8 - 9 | 927 | 72 | ||
NR_049785.1 | 6 - 7 | 860 | 72 | ||
NR_049786.1 | 7 - 8 | 809 | 72 | ||
NR_049787.1 | 5 - 6 | 660 | 72 | ||
XM_011533203.1 | XP_011531505.1 | 6 - 7 | 627 | 72 | |
XM_017005405.1 | XP_016860894.1 | 5 - 6 | 797 | 72 | |
GenBank mRNA | AK129689.1 | - | 7 - 8 | 737 | 72 |
AK222543.1 | - | 7 - 8 | 736 | 72 | |
AK293464.1 | - | 7 - 8 | 669 | 72 | |
AK295278.1 | - | 5 - 6 | 539 | 72 | |
AK297054.1 | - | 7 - 8 | 862 | 72 | |
AK298620.1 | - | 7 - 8 | 688 | 72 | |
AK298856.1 | - | 7 - 8 | 690 | 72 | |
AK298869.1 | - | 6 - 7 | 639 | 72 | |
AK300144.1 | - | 7 - 8 | 700 | 72 | |
AK300641.1 | - | 5 - 6 | 531 | 72 | |
AK307122.1 | - | 6 - 7 | 736 | 72 | |
AK308487.1 | - | 6 - 7 | 739 | 72 | |
AK309640.1 | - | 5 - 6 | 781 | 72 | |
AK316373.1 | - | 7 - 8 | 632 | 72 | |
BC002524.1 | - | 7 - 8 | 714 | 72 | |
BC002610.1 | - | 7 - 8 | 703 | 72 | |
BC014620.2 | - | 7 - 8 | 714 | 72 | |
BC050590.1 | - | 7 - 8 | 708 | 72 | |
BT007167.1 | - | 7 - 8 | 605 | 72 | |
CR457081.1 | - | 7 - 8 | 605 | 72 | |
D31764.1 | - | 7 - 8 | 827 | 72 |