The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172222.2 | NP_001165693.1 | 3 - 4 | 306 | 164 |
NM_012236.3 | NP_036368.1 | 5 - 6 | 521 | 164 | |
XM_011541035.2 | XP_011539337.1 | 5 - 6 | 531 | 164 | |
XM_017000700.1 | XP_016856189.1 | 5 - 6 | 532 | 164 | |
XM_017000703.1 | XP_016856192.1 | 5 - 6 | 531 | 164 | |
XM_017000705.1 | XP_016856194.1 | 6 - 7 | 732 | 164 | |
XM_017000707.1 | XP_016856196.1 | 6 - 7 | 732 | 164 | |
GenBank mRNA | AK299383.1 | - | 1 - 2 | 151 | 164 |
BX420965.2 | - | 4 - 5 | 439 | 164 |