Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCMH1
Entrez Gene ID
22955
Gene Name
sex comb on midleg homolog 1 (Drosophila)
Gene Aliases
Scml3
Chromosome Location
Chr.1: 41027199 - 41242155 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031694.2 NP_001026864.1 8 - 9 1083 71
NM_001172218.1 NP_001165689.1 9 - 10 1155 71
NM_001172219.1 NP_001165690.1 8 - 9 946 71
NM_001172220.1 NP_001165691.1 7 - 8 875 71
NM_001172221.1 NP_001165692.1 7 - 8 970 71
NM_012236.3 NP_036368.1 8 - 9 1084 71
NR_135092.1 8 - 9 1083 71
XM_006710462.2 XP_006710525.1 8 - 9 946 71
XM_006710464.1 XP_006710527.1 7 - 8 895 71
XM_011541032.2 XP_011539334.1 8 - 9 884 71
XM_011541033.2 XP_011539335.1 10 - 11 1301 71
XM_011541034.1 XP_011539336.1 8 - 9 946 71
XM_011541035.2 XP_011539337.1 8 - 9 1094 71
XM_011541036.2 XP_011539338.1 9 - 10 1157 71
XM_011541040.2 XP_011539342.1 8 - 9 946 71
XM_011541043.2 XP_011539345.1 10 - 11 1397 71
XM_011541044.1 XP_011539346.1 3 - 4 513 71
XM_011541045.2 XP_011539347.2 9 - 10 1170 71
XM_017000698.1 XP_016856187.1 5 - 6 699 71
XM_017000699.1 XP_016856188.1 7 - 8 885 71
XM_017000700.1 XP_016856189.1 8 - 9 1095 71
XM_017000702.1 XP_016856191.1 8 - 9 960 71
XM_017000703.1 XP_016856192.1 8 - 9 1098 71
XM_017000709.1 XP_016856198.1 7 - 8 922 71
GenBank mRNA AF149045.1 - 8 - 9 1057 71
AF149046.1 - 9 - 10 1128 71
AK023124.1 - 8 - 9 913 71
AK301497.1 - 6 - 7 925 71
BC009752.2 - 7 - 8 1025 71
BC021252.2 - 7 - 8 983 71
BX640721.1 - 7 - 8 861 71
CR457161.1 - 3 - 4 531 71

View Full Product Details