The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198185.4 | NP_937828.3 | 14 - 15 | 150 | |
XM_017017659.1 | XP_016873148.1 | 14 - 15 | 150 | ||
GenBank mRNA | AW271430.1 | - | 2 - 3 | 252 | 150 |