The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145368.1 | NP_001138840.1 | 11 - 12 | 942 | 150 |
NM_001145369.1 | NP_001138841.1 | 6 - 7 | 2991 | 150 | |
NM_001145370.1 | NP_001138842.1 | 6 - 7 | 2991 | 150 | |
NM_002829.3 | NP_002820.3 | 11 - 12 | 942 | 150 | |
XM_006717197.3 | XP_006717260.1 | 11 - 12 | 1051 | 150 | |
XM_006717199.3 | XP_006717262.1 | 11 - 12 | 1076 | 150 | |
XM_006717201.3 | XP_006717264.1 | 10 - 11 | 883 | 150 | |
XM_006717202.3 | XP_006717265.1 | 8 - 9 | 645 | 150 | |
XM_006717203.3 | XP_006717266.1 | 8 - 9 | 604 | 150 | |
XM_006717204.3 | XP_006717267.1 | 11 - 12 | 978 | 150 | |
XM_011518888.2 | XP_011517190.1 | 11 - 12 | 1012 | 150 | |
XM_011518889.2 | XP_011517191.1 | 11 - 12 | 1682 | 150 | |
XM_017014955.1 | XP_016870444.1 | 11 - 12 | 1034 | 150 | |
XM_017014956.1 | XP_016870445.1 | 10 - 11 | 890 | 150 | |
XM_017014957.1 | XP_016870446.1 | 11 - 12 | 978 | 150 | |
GenBank mRNA | AK295875.1 | - | 10 - 11 | 862 | 150 |
AK304434.1 | - | 11 - 12 | 942 | 150 | |
AK316066.1 | - | 11 - 12 | 942 | 150 | |
BC126117.1 | - | 10 - 11 | 852 | 150 | |
BC143848.1 | - | 10 - 11 | 852 | 150 | |
BX648735.1 | - | 6 - 7 | 1426 | 150 | |
M64572.1 | - | 10 - 11 | 853 | 150 | |
S39392.1 | - | 3 - 4 | 253 | 150 |