The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145369.1 | NP_001138841.1 | 7 - 8 | 3157 | 118 |
NM_001145371.1 | NP_001138843.1 | 2 - 3 | 591 | 118 | |
NM_002829.3 | NP_002820.3 | 12 - 13 | 1108 | 118 | |
XM_006717197.3 | XP_006717260.1 | 12 - 13 | 1217 | 118 | |
XM_006717199.3 | XP_006717262.1 | 12 - 13 | 1242 | 118 | |
XM_006717201.3 | XP_006717264.1 | 11 - 12 | 1049 | 118 | |
XM_006717202.3 | XP_006717265.1 | 9 - 10 | 811 | 118 | |
XM_006717203.3 | XP_006717266.1 | 9 - 10 | 770 | 118 | |
XM_006717204.3 | XP_006717267.1 | 12 - 13 | 1144 | 118 | |
XM_011518888.2 | XP_011517190.1 | 12 - 13 | 1178 | 118 | |
XM_011518889.2 | XP_011517191.1 | 12 - 13 | 1848 | 118 | |
XM_017014955.1 | XP_016870444.1 | 12 - 13 | 1200 | 118 | |
GenBank mRNA | AK296388.1 | - | 2 - 3 | 517 | 118 |
BC126117.1 | - | 11 - 12 | 1018 | 118 | |
BX648735.1 | - | 7 - 8 | 1592 | 118 | |
DQ104439.1 | - | 11 - 12 | 996 | 118 | |
M64572.1 | - | 11 - 12 | 1019 | 118 | |
S39392.1 | - | 4 - 5 | 419 | 118 |