Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMOD1
Entrez Gene ID
7111
Gene Name
tropomodulin 1
Gene Aliases
D9S57E, ETMOD, TMOD
Chromosome Location
Chr.9: 97501180 - 97601743 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166116.1 NP_001159588.1 9 - 10 1154 75
NM_003275.3 NP_003266.1 9 - 10 1231 75
GenBank mRNA AK095748.1 - 6 - 7 898 75
AK314533.1 - 10 - 11 1154 75
BC002660.2 - 9 - 10 1231 75
M77016.1 - 8 - 9 1052 75

View Full Product Details