Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMOD1
Entrez Gene ID
7111
Gene Name
tropomodulin 1
Gene Aliases
D9S57E, ETMOD, TMOD
Chromosome Location
Chr.9: 97501180 - 97601743 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166116.1 NP_001159588.1 2 - 3 257 84
NM_003275.3 NP_003266.1 2 - 3 334 84
GenBank mRNA AK314533.1 - 2 - 3 257 84
BC002660.2 - 2 - 3 334 84
DA015088.1 - 2 - 3 307 84
M77016.1 - 1 - 2 155 84

View Full Product Details