Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMOD1
Entrez Gene ID
7111
Gene Name
tropomodulin 1
Gene Aliases
D9S57E, ETMOD, TMOD
Chromosome Location
Chr.9: 97501180 - 97601743 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166116.1 NP_001159588.1 3 - 4 414 69
NM_003275.3 NP_003266.1 3 - 4 491 69
GenBank mRNA AK314533.1 - 3 - 4 414 69
BC002660.2 - 3 - 4 491 69
DA015088.1 - 3 - 4 464 69
M77016.1 - 2 - 3 312 69

View Full Product Details