The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166116.1 | NP_001159588.1 | 5 - 6 | 618 | 64 |
NM_003275.3 | NP_003266.1 | 5 - 6 | 695 | 64 | |
GenBank mRNA | AK095748.1 | - | 2 - 3 | 362 | 64 |
AK314533.1 | - | 5 - 6 | 618 | 64 | |
BC002660.2 | - | 5 - 6 | 695 | 64 | |
M77016.1 | - | 4 - 5 | 516 | 64 |