Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FGF14
- Entrez Gene ID
-
2259
- Gene Name
- fibroblast growth factor 14
- Gene Aliases
-
FGF-14, FHF-4, FHF4, SCA27
- Chromosome Location
-
Chr.13: 101720855 - 102402428 on Build GRCh38
View Full Product Details