The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004501.3 | NP_004492.2 | 2 - 3 | 968 | 101 |
NM_031844.2 | NP_114032.2 | 2 - 3 | 1025 | 101 | |
XM_017001115.1 | XP_016856604.1 | 2 - 3 | 1220 | 101 | |
XM_017001116.1 | XP_016856605.1 | 2 - 3 | 1025 | 101 | |
XM_017001117.1 | XP_016856606.1 | 2 - 3 | 1014 | 101 | |
GenBank mRNA | AF068846.1 | - | 2 - 3 | 1021 | 101 |
AK126868.1 | - | 3 - 4 | 766 | 101 | |
AK130669.1 | - | 1 - 2 | 74 | 101 | |
AK297119.1 | - | 3 - 4 | 859 | 101 | |
AK307661.1 | - | 2 - 3 | 492 | 101 | |
BC003367.1 | - | 2 - 3 | 899 | 101 | |
BC003621.2 | - | 2 - 3 | 901 | 101 | |
BC007950.2 | - | 2 - 3 | 449 | 101 | |
BC015782.1 | - | 2 - 3 | 502 | 101 | |
BC024767.1 | - | 2 - 3 | 901 | 101 | |
X65488.1 | - | 2 - 3 | 791 | 101 |