The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000136.2 | NP_000127.2 | 14 - 15 | 1798 | 71 |
NM_001243743.1 | NP_001230672.1 | 14 - 15 | 1740 | 71 | |
XM_005251802.3 | XP_005251859.1 | 8 - 9 | 1900 | 71 | |
XM_006717001.2 | XP_006717064.1 | 13 - 14 | 1615 | 71 | |
XM_011518365.2 | XP_011516667.1 | 14 - 15 | 1773 | 71 | |
XM_011518367.2 | XP_011516669.1 | 15 - 16 | 1910 | 71 | |
XM_017014452.1 | XP_016869941.1 | 15 - 16 | 1925 | 71 | |
XM_017014453.1 | XP_016869942.1 | 15 - 16 | 1872 | 71 | |
XM_017014454.1 | XP_016869943.1 | 14 - 15 | 1799 | 71 | |
GenBank mRNA | AK312548.1 | - | 14 - 15 | 1663 | 71 |
BC015748.1 | - | 14 - 15 | 1788 | 71 | |
CB052780.1 | - | 1 - 2 | 101 | 71 | |
KU178051.1 | - | 13 - 14 | 1536 | 71 | |
KU178052.1 | - | 14 - 15 | 1681 | 71 | |
X66893.1 | - | 14 - 15 | 1788 | 71 | |
X66894.1 | - | 14 - 15 | 1706 | 71 |