The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098525.2 | NP_001091995.1 | 4 - 5 | 1304 | 60 |
NM_001286686.1 | NP_001273615.1 | 4 - 5 | 1242 | 60 | |
NM_001286687.1 | NP_001273616.1 | 4 - 5 | 1301 | 60 | |
NM_018204.4 | NP_060674.3 | 4 - 5 | 1301 | 60 | |
XM_005266344.4 | XP_005266401.1 | 4 - 5 | 1212 | 60 | |
XM_011535043.2 | XP_011533345.1 | 3 - 4 | 1156 | 60 | |
GenBank mRNA | AF177227.1 | - | 3 - 4 | 996 | 60 |
AK001611.1 | - | 1 - 2 | 177 | 60 | |
AK022982.1 | - | 4 - 5 | 1200 | 60 | |
AK299083.1 | - | 4 - 5 | 1242 | 60 | |
AL136848.1 | - | 1 - 2 | 706 | 60 | |
AY062261.1 | - | 4 - 5 | 1222 | 60 | |
AY062262.1 | - | 4 - 5 | 1222 | 60 | |
BC010901.1 | - | 1 - 2 | 462 | 60 | |
BC130296.1 | - | 4 - 5 | 1149 | 60 | |
BC136332.1 | - | 4 - 5 | 1149 | 60 | |
EF560732.1 | - | 4 - 5 | 1185 | 60 | |
Y15758.1 | - | 4 - 5 | 1197 | 60 |