Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLPTM1L
Entrez Gene ID
81037
Gene Name
CLPTM1 like
Gene Aliases
CRR9
Chromosome Location
Chr.5: 1317744 - 1345070 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030782.4 NP_110409.2 3 - 4 682 81
GenBank mRNA AB045223.1 - 3 - 4 439 81
AK027306.1 - 3 - 4 496 81
BC016399.1 - 1 - 2 68 81
BC025305.1 - 3 - 4 500 81

View Full Product Details