The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203315.2 | NP_976060.1 | 1 - 2 | 148 | 78 |
GenBank mRNA | AV645953.1 | - | 1 - 2 | 139 | 78 |
H80183.1 | - | 1 - 2 | 147 | 77 |