Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TARBP1
Entrez Gene ID
6894
Gene Name
TAR (HIV-1) RNA binding protein 1
Gene Aliases
TRM3, TRP-185, TRP185
Chromosome Location
Chr.1: 234391313 - 234479174 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005646.3 NP_005637.3 14 - 15 2391 145
XM_005273234.3 XP_005273291.1 14 - 15 2388 145
XM_011544263.2 XP_011542565.1 14 - 15 2458 145
XM_017002194.1 XP_016857683.1 14 - 15 2461 145
XM_017002195.1 XP_016857684.1 11 - 12 1264 145
XM_017002196.1 XP_016857685.1 2 - 3 236 145
GenBank mRNA U38847.1 - 14 - 15 2391 145

View Full Product Details