Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TARBP1
Entrez Gene ID
6894
Gene Name
TAR (HIV-1) RNA binding protein 1
Gene Aliases
TRM3, TRP-185, TRP185
Chromosome Location
Chr.1: 234391313 - 234479174 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005646.3 NP_005637.3 26 - 27 4239 152
XM_005273234.3 XP_005273291.1 26 - 27 4236 152
XM_011544263.2 XP_011542565.1 25 - 26 4234 152
XM_017002195.1 XP_016857684.1 23 - 24 3112 152
XM_017002196.1 XP_016857685.1 14 - 15 2084 152
XM_017002197.1 XP_016857686.1 21 - 22 2852 152
GenBank mRNA BC016023.1 - 4 - 5 828 152
U38847.1 - 26 - 27 4239 152

View Full Product Details