Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KSR2
Entrez Gene ID
283455
Gene Name
kinase suppressor of ras 2
Gene Aliases
-
Chromosome Location
Chr.12: 117453012 - 117968558 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173598.4 NP_775869.3 16 - 17 2417 73
XM_011538224.2 XP_011536526.1 16 - 17 2746 73
XM_011538225.2 XP_011536527.1 16 - 17 3211 73
XM_011538228.2 XP_011536530.1 14 - 15 1576 73
XM_017019208.1 XP_016874697.1 16 - 17 2752 73
XM_017019210.1 XP_016874699.1 11 - 12 1202 73
GenBank mRNA AY345972.1 - 14 - 15 2086 73
BC107106.1 - 14 - 15 2081 73
BC107107.1 - 14 - 15 2081 73
BC127603.1 - 14 - 15 2086 73

View Full Product Details